EC900

Sigma-Aldrich

GenElute-E FFPE Deparaffinization Solution

greener alternative
价格与库存信息目前不能提供

purified by

(Time: About 10 minutes)
(Xylene alternative)

feature

Intended use (For deparaffinization of FFPE samples)

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

greener alternative category

Aligned

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is aligned with "Waste prevention" and "Designing safer chemicals". Click here for more information.
For deparaffinization of up to four slices of FFPE sample per purification to be used with silica nucleic acid isolation kits for isolation of DNA and/or RNA.

Application

Traditional methods for deparaffinaization of FFPE sample for DNA or RNA isolation rely on hazardous chemicals such as xylene.GenElute-E FFPE Deparaffinization Solution provides a safer alternative, helping labs to meet sustainability or waste-reduction goals.

Features and Benefits

Safer alternative is hazardous chemicals such as xylene

Other Notes

We believe that green chemistry will contribute to a better tomorrow. With a growing portfolio of greener alternatives, there are now more choices to reduce the ecological impact of your research while still delivering quality and efficacy so your results are not compromised. GenElute-E kits adhere to the principles of SMASH Packaging, our global initiative to improve the sustainability through the reduction of packaging materials, the increased use of sustainable alternatives, and improved recyclability. Packaging is FCS-marked for sustainable forestry practices and produced from greater than 70% recycled content.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Supp Hazards

EUH066

WGK Germany

WGK 1

Flash Point(F)

275.0 °F

Flash Point(C)

135 °C

技术文章
Impact of Purification Method on Accuracy of DNA Quantitation and Downstream Enzymatic Processes. Evaluation of the purity of genomic DNA by UV spectrophotometry, gel electrophoresis, and downstream qPCR using GenElute™-E DNA purification kits.
了解更多
相关内容
Reduce plastic waste and eliminate hazardous liquid waste for more sustainable laboratories with GenElute™-E Single Spin DNA and RNA prep kits.
了解更多
Demo video showing how to purify DNA and RNA GenElute™-E single spin nucleic acid purification kits
了解更多
Animation showing technology principle of GenElute™-E single spin negative chromatography nucleic acid purification kits
了解更多

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。