MAB2141Z

Sigma-Aldrich

抗整合素α2β 1抗体,克隆BMA2.1,不含叠氮化物

clone BMA2.1, Chemicon®, from rat

别名:
VLA-2, MAB2141
eCl@ss:
32160702
NACRES:
NA.41
价格与库存信息目前不能提供

Quality Level

biological source

rat

antibody product type

primary antibodies

clone

BMA2.1, monoclonal

species reactivity

mouse

mfr. no.

Chemicon®

application(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... ITGA2(3673)

Specificity

抗小鼠整联素 α2β1 异二聚体。 抗体不能单独识别 α2 或 β1 亚基。

Immunogen

小鼠整合素 α2β1 异二聚体

Application

使用该抗整合素 α2β1 抗体(克隆 BMA2.1,无叠氮化物,经验证可用于 FC & IP)检测整合素 α2β1。
研究子类别
整合素
流式细胞术

阻断整联素 α2β1 介导的粘附作用

免疫沉淀

对免疫组化无效。

最佳工作稀释度必须由最终用户确定。
研究类别
细胞结构

Physical form

不含防腐剂的液体。
形式:纯化的

Storage and Stability

未稀释的等分试样保持在 2-8°C 下可保存长达 12 个月。

Other Notes

浓度:请参考批次特异性浓缩物的分析证书。

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

除非我们的产品目录或产品附带的其他公司文档另有说明,否则我们的产品仅供研究使用,不得用于任何其他目的,包括但不限于未经授权的商业用途、体外诊断用途、离体或体内治疗用途或任何类型的消费或应用于人类或动物。
分析证书
原产地证书 (CofO)
Steffen Massberg et al.
The Journal of experimental medicine, 203(5), 1221-1233 (2006-04-19)
The accumulation of smooth muscle and endothelial cells is essential for remodeling and repair of injured blood vessel walls. Bone marrow-derived progenitor cells have been implicated in vascular repair and remodeling; however, the mechanisms underlying their recruitment to the site...
Mubeen Jafri et al.
American journal of physiology. Gastrointestinal and liver physiology, 295(1), G16-G26 (2008-04-26)
Inoculation of BALB/c mice with rhesus rotavirus (RRV) in the newborn period results in biliary epithelial cell (cholangiocyte) infection and the murine model of biliary atresia. Rotavirus infection of a cell requires attachment, which is governed in part by cell-surface...
Yuan Cao et al.
Cancer research, 74(18), 5091-5102 (2014-09-17)
The TGFβ growth factor family member BMP4 is a potent suppressor of breast cancer metastasis. In the mouse, the development of highly metastatic mammary tumors is associated with an accumulation of myeloid-derived suppressor cells (MDSC), the numbers of which are...

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。