Merck
All Photos(1)

R03040

Sigma-Aldrich

Eosin Stain Solution, 5% Aqueous

Cytoplasmic stain, H&E, 5% aqueous

Synonym(s):
Counterstain

质量水平

形式

liquid

mp

-0.1 °C

转变温度

flash point 36 °C

λmax

515 nm

application(s)

diagnostic assay manufacturing
hematology
histology

运输

ambient

储存温度

15-25°C

Compare Similar Items

View Full Comparison

1 of 4

This Item
592-M1.005911.02439
form

liquid

form

liquid

form

liquid

form

liquid

mp

-0.1 °C

mp

-0.1 °C

mp

-

mp

-

λmax

515 nm

λmax

510-520 nm

λmax

-

λmax

-

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology, histology

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

shipped in

-

shipped in

-

shipped in

-

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

2-8°C

storage temp.

15-25°C

应用

Cytoplasmic stain, H&E, 5% aqueous
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.

象形图

FlameExclamation mark

警示用语:

Warning

危险声明

危险分类

Eye Irrit. 2 - Flam. Liq. 3 - Skin Sens. 1

储存分类代码

3 - Flammable liquids

WGK

WGK 3

闪点(F)

96.8 °F

闪点(C)

36 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

(enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service