Merck

0958A-BM

Sigma-Aldrich

GlcNAcβ3`LN-C3-biot

登录查看公司和协议定价

偶联物

biotin conjugate

产品线

BM-Series

检测方案

95% (H-NMR)

形式

lyophilized powder

分子量

983.1 Da

manufacturer/tradename

GlycoNZ

标记范围

20 mol% Biotin ligand density

technique(s)

glycol-bead construction: suitable
glycol-surface construction: suitable
microarray: suitable

probe type

trisaccharide

probe presentation

monomeric

储存温度

−20°C

相关类别

比较类似商品

查看完整比较结果

显示差异

1 of 4

此商品
0864-BM0075-BM0011-BM
vibrant-m

0958A-BM

GlcNAcβ3`LN-C3-biot

vibrant-m

0864-BM

GlcNAcα3`LN-C3-biot

vibrant-m

0075-BM

GlcNAcβ3`TF-C3-biot

vibrant-m

0011-BM

GlcNAcβ2`TF-C3-biot

conjugate

biotin conjugate

conjugate

biotin conjugate

conjugate

biotin conjugate

conjugate

biotin conjugate

technique(s)

glycol-bead construction: suitable, microarray: suitable, glycol-surface construction: suitable

technique(s)

glycol-bead construction: suitable, glycol-surface construction: suitable, microarray: suitable

technique(s)

glycol-bead construction: suitable, glycol-surface construction: suitable, microarray: suitable

technique(s)

glycol-bead construction: suitable, glycol-surface construction: suitable, microarray: suitable

manufacturer/tradename

GlycoNZ

manufacturer/tradename

GlycoNZ

manufacturer/tradename

GlycoNZ

manufacturer/tradename

GlycoNZ

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

extent of labeling

20 mol% Biotin ligand density

extent of labeling

20 mol% Biotin ligand density

extent of labeling

20 mol% Biotin ligand density

extent of labeling

20 mol% Biotin ligand density

一般描述

BM-Series: Biotinylated Monomeric Probes
These glycoconjugate probes have an elongated spacer giving them the possibility to bind avidin or streptavidin. The biotin fragment is hydrophobic enough to permit the probes to be adsorbed on SepPack C18 cartridges from aqueous solutions, and to be eluted by methanol, similarly to glycosides bearing a “Lemieux-type” space. The spacer is the normal spacer of the ligand plus the spacer of the biotin label –CO(CH2)5NH–.

应用

Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.

储存分类代码

13 - Non Combustible Solids

WGK

WGK 3

闪点(°F)

Not applicable

闪点(°C)

Not applicable


分析证书(COA)

输入产品批号来搜索 分析证书(COA) 。批号可以在产品标签上"批“ (Lot或Batch)字后找到。

已有该产品?

为方便起见,与您过往购买产品相关的文件已保存在文档库中。

访问文档库

难以找到您所需的产品或批次号码?

在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).

示例

T1503
货号
-
25G
包装规格/数量

其它示例:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

输入内容 1.000309185)

遇到问题?欢迎随时联系我们技术服务 寻求帮助

批号可以在产品标签上"批“ (Lot或Batch)字后面找到。

Aldrich 产品

  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。

  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。

  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。

未找到您寻找的产品?

部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。

索取COA

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门