Merck
All Photos(2)

0980-PA

Sigma-Aldrich

LN6TF-C3-PAA

carrier

PAA carrier

产品线

PA-Series

检测方案

95% (H-NMR)

形式

lyophilized powder

分子量

~20,000 Da

manufacturer/tradename

GlycoNZ

标记范围

20 mol% Poly[N-(2-hydroxyethyl)acrylamide-co-N-acrylamide] ligand density

technique(s)

ELISA: suitable

probe type

tetrasaccharide

probe presentation

polymeric

储存温度

−20°C

Related Categories

Compare Similar Items

View Full Comparison

1 of 4

This Item
0980-BP0980-FP0871-PA
LN6TF-C3-PAA

Sigma-Aldrich

0980-PA

LN6TF-C3-PAA

LN6TF-C3-PAA-biot

Sigma-Aldrich

0980-BP

LN6TF-C3-PAA-biot

LN6TF-C3-PAA-fluo

Sigma-Aldrich

0980-FP

LN6TF-C3-PAA-fluo

LN6`LN-C2-PAA

Sigma-Aldrich

0871-PA

LN6`LN-C2-PAA

carrier

PAA carrier

carrier

PAA carrier

carrier

PAA carrier

carrier

PAA carrier

product line

PA-Series

product line

BP-Series

product line

FP-Series

product line

PA-Series

assay

95% (H-NMR)

assay

95% (H-NMR)

assay

95% (H-NMR)

assay

95% (H-NMR)

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

mol wt

~20,000 Da

mol wt

~20,000 Da

mol wt

~20,000 Da

mol wt

~20,000 Da

manufacturer/tradename

GlycoNZ

manufacturer/tradename

GlycoNZ

manufacturer/tradename

GlycoNZ

manufacturer/tradename

GlycoNZ

一般描述

PA-Series: Unlabeled Polyacrylamide Conjugates
This series of glycoconjugate probes consists of HOCH2CH2NH3+-salts for carboxylates and sulfates. PAA is poly[N-(2-hydroxyethyl) acrylamide], a flexible polymer chain which serves as an additional spacer. The spacer-arm for oligosaccharides is normally –(CH2)3–.

应用

Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.

制备说明

Purity: synthetic ω-aminoalkyl glycosides used for coupling with the polymer have a purity of >95% (HPLC and 1H-NMR)

储存分类代码

13 - Non Combustible Solids

WGK

WGK 3

闪点(F)

Not applicable

闪点(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

(enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service