About This Item
分類程式碼代碼:
12352200
技术服务
需要帮助?我们经验丰富的科学家团队随时乐意为您服务。
让我们为您提供帮助
形狀
solution
包裝
pkg of 5 × 10 mL
一般說明
This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of rubidium. A unit of SRM 3145a consists of five 10 mL sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically to contain a known mass fraction of rubidium. The solution contains nitric acid at a volume fraction of approximately 1 %, which is equivalent to an amount-of-substance concentration (molarity) of approximately 0.16 mol/L. SRM 3145A_cert SRM 3145A _SDS
法律資訊
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
历史批次信息供参考:
分析证书(COA)