形狀
solution
包裝
pkg of 5 × 10 mL
一般說明
This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of rubidium. A unit of SRM 3145a consists of five 10 mL sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically to contain a known mass fraction of rubidium. The solution contains nitric acid at a volume fraction of approximately 1 %, which is equivalent to an amount-of-substance concentration (molarity) of approximately 0.16 mol/L. SRM 3145A_cert SRM 3145A _SDS
法律資訊
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology