1 of 4
This Item | PEA1910 | PEA1912 | PEA1911 |
---|---|---|---|
application(s) environmental | application(s) environmental | application(s) environmental | application(s) environmental |
format matrix material | format matrix material | format matrix material | format matrix material |
agency EPA (Method 12) | agency EPA (Method 29) | agency EPA (Method 29) | agency EPA (Method 13, 14) |
packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
13 - Non Combustible Solids
WGK 3
Not applicable
Not applicable
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service