Merck
All Photos(1)

SK002

Supelco

PCB CALIBRATION VERIFICATION 2

质量水平

100

等级

certified reference material

格式

multi-component solution

储存温度

15-25°C

一般描述

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    Aroclor-1016 (PCB-1016) 0.1 µg/mL
    Aroclor-1260 (PCB-1260) 0.1 µg/mL
    PCBs as Decachlorobiphenyl 0.005 µg/mL

警示用语:

Danger

危险分类

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

靶器官

Central nervous system

储存分类代码

3 - Flammable liquids

WGK

WGK 3

闪点(F)

10.4 °F - closed cup

闪点(C)

-11.99 °C - closed cup

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service