Merck

107S

Sigma-Aldrich

CD7 Positive Control Slides

suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections)

NACRES:
NA.24

包装

pkg of 5 slides

质量水平

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

运输

ambient

储存温度

no temp limit

比较类似商品

查看完整比较结果

Show Differences

1 of 4

此商品
161S263S171S
Sigma-Aldrich

Sigma-Aldrich

107S

CD7 Positive Control Slides

Sigma-Aldrich

Sigma-Aldrich

161S

CD61 Positive Control Slides

Sigma-Aldrich

Sigma-Aldrich

263S

CD63 Positive Control Slides

Sigma-Aldrich

Sigma-Aldrich

171S

CD71 Positive Control Slides

manufacturer/tradename

Cell Marque

manufacturer/tradename

Cell Marque

manufacturer/tradename

Cell Marque

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

no temp limit

Quality Level

100

Quality Level

100, 500

Quality Level

100, 500

Quality Level

100, 500

一般描述

CD7 positive control slides are intended for use as positive controls for immunohistochemical (IHC) staining using an anti-CD7 antibody.

外形

Paraffin embedded tissue section mounted on microscope slide

其他说明

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

法律信息

Cell Marque is a trademark of Sigma-Aldrich Co. LLC

抗体

产品编号
说明
价格

储存分类代码

11 - Combustible Solids

WGK

WGK 3

闪点(F)

Not applicable

闪点(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例 T1503).

示例

T1503
货号
-
25G
包装规格/数量

其它示例:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

请输入1.000309185)

遇到问题?欢迎随时联系我们技术服务 寻求帮助

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich 产品

  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。

  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。

  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。

未找到您寻找的产品?

部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。

索取COA

Ana-Marija Jagatić Korenika et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-26)
Commercial sparkling wine production represents a relatively low but important part of the Croatian wine production, especially in the Zagreb county. This study presents the results of volatile aroma compounds profile and organic acid composition of commercial sparkling wine samples

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门