Merck
All Photos(1)

117S

Sigma-Aldrich

CD117, c-kit Positive Control Slides

suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections)

NACRES:
NA.24

包装

pkg of 5 slides

质量水平

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

运输

ambient

储存温度

no temp limit

Compare Similar Items

View Full Comparison

1 of 4

This Item
156S198S123S
packaging

pkg of 5 slides

packaging

pkg of 5 slides

packaging

pkg of 5 slides

packaging

pkg of 5 slides

manufacturer/tradename

Cell Marque

manufacturer/tradename

Cell Marque

manufacturer/tradename

Cell Marque

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

no temp limit

Quality Level

100

Quality Level

100, 500

Quality Level

100, 500

Quality Level

100, 500

一般描述

CD117, c-kit positive control slides are intended for use as positive controls for immunohistochemical (IHC) staining using an anti-CD117, c-kit antibody.

外形

Paraffin embedded tissue section mounted on microscope slide

其他说明

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

法律信息

Cell Marque is a trademark of Sigma-Aldrich Co. LLC

储存分类代码

13 - Non Combustible Solids

WGK

WGK 3

闪点(F)

Not applicable

闪点(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

(enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service