Merck

APREST79483

Sigma-Aldrich

PrEST Antigen GFM1

Prestige Antigens Powered by Atlas Antibodies, buffered aqueous solution

登录to View Organizational & Contract Pricing

别名:
EFGM, EGF1, GFM

重组

expressed in E. coli

质量水平

检测方案

>80% (SDS-PAGE)

形式

buffered aqueous solution

分子量

predicted mol wt 27 kDa

纯化方式

immobilized metal affinity chromatography (IMAC)

浓度

≥0.5 mg/mL

免疫原序列

QYGKVIGVLEPLDPEDYTKLEFSDETFGSNIPKQFVPAVEKGFLDACEKGPLSGHKLSGLRFVLQDGAHHMVDSNEISFIRAGE

基因组数据库 |人类登记号

UniProt登记号

运输

wet ice

储存温度

−20°C

Gene Information

human ... GFM1(85476)

一般描述

Recombinant protein fragment of Human GFM1 with N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G) fusion tag.

应用

Suitable as a blocking agent using corresponding antibodies.

联系

Corresponding Antibody HPA034765.

外形

Solution in 1 M urea-PBS, pH 7.4

制备说明

The protein solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user.

法律信息

Prestige Antigens is a trademark of Sigma-Aldrich Co. LLC

免责声明

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

储存分类代码

10 - Combustible liquids

WGK

WGK 2

闪点(°F)

Not applicable

闪点(°C)

Not applicable


分析证书(COA)

输入产品批号来搜索 分析证书(COA) 。批号可以在产品标签上"批“ (Lot或Batch)字后找到。

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).

示例

T1503
货号
-
25G
包装规格/数量

其它示例:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

输入内容 1.000309185)

遇到问题?欢迎随时联系我们技术服务 寻求帮助

批号可以在产品标签上"批“ (Lot或Batch)字后面找到。

Aldrich 产品

  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。

  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。

  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。

未找到您寻找的产品?

部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。

索取COA

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门