AV13027

Sigma-Aldrich

Anti-GABRA3 (AB2) antibody produced in rabbit

IgG fraction of antiserum

NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

rabbit, rat, human, guinea pig, dog, bovine, mouse, horse

concentration

0.5 mg - 1 mg/mL

application(s)

immunohistochemistry: suitable
western blot: suitable

conjugate

unconjugated

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GABRA3(2556)

Immunogen

Synthetic peptide directed towards the N terminal region of human GABRA3

Application

Anti-GABRA3 (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml.

Biochem/physiol Actions

GABRA3 is a subunit of GABAA receptor, the main inhibitory receptor in the brain. This subunit is mainly targeted to mouse amygdala, Purkinje cells and bulbus olfactorius.

Sequence

Synthetic peptide located within the following region: GTTGQGESRRQEPGDFVKQDIGGLSPKHAPDIPDDSTDNITIFTRILDRL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

分析证书
原产地证书 (CofO)
H Hörtnagl et al.
Neuroscience, 236, 345-372 (2013-01-23)
The GABAA receptor is the main inhibitory receptor in the brain and its subunits originate from different genes or gene families (α1-α6, β1-β3, γ1-γ3, δ, ε, θ, π, or ρ1-3). In the mouse brain the anatomical distribution of GABAA receptor...

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。