AV32368

Sigma-Aldrich

Anti-GLI1 antibody produced in rabbit

IgG fraction of antiserum

别名:
Anti-Glioma-associated oncogene homolog 1 (zinc finger protein)
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

118 kDa

species reactivity

dog, human, rabbit, guinea pig, horse, rat, bovine

concentration

0.5 mg - 1 mg/mL

application(s)

western blot: suitable

conjugate

unconjugated

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GLI1(2735)

Immunogen

Synthetic peptide directed towards the C terminal region of human GLI1

Biochem/physiol Actions

GLI1 is a protein which is a member of the Kruppel family of zinc finger proteins. The function of this gene has not been determined; however, it may play a role in normal development gene transcription. Mouse mutation studies indicate possible involvement in human foregut malformation.

Sequence

Synthetic peptide located within the following region: TNPSCGHPEVGRLGGGPALYPPPEGQVCNPLDSLDLDNTQLDFVAILDEP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

分析证书
原产地证书 (CofO)
Young-Eun Leem et al.
PloS one, 9(11), e111701-e111701 (2014-11-05)
Hedgehog (Hh) signaling plays essential roles in various developmental processes, and its aberrant regulation results in genetic disorders or malignancies in various tissues. Hyperactivation of Hh signaling is associated with lung cancer development, and there have been extensive efforts to...
Yoshiaki Kitaura et al.
PloS one, 9(10), e109597-e109597 (2014-10-15)
Hedgehog (Hh) signaling plays important roles in various development processes. This signaling is necessary for osteoblast formation during endochondral ossification. In contrast to the established roles of Hh signaling in embryonic bone formation, evidence of its roles in adult bone...
Ann Marie Hynes et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(27), 9893-9898 (2014-06-21)
Nephronophthisis (NPHP) is the major cause of pediatric renal failure, yet the disease remains poorly understood, partly due to the lack of appropriate animal models. Joubert syndrome (JBTS) is an inherited ciliopathy giving rise to NPHP with cerebellar vermis aplasia...
Natsuko Kageyama-Yahara et al.
PloS one, 9(8), e106106-e106106 (2014-08-29)
MUC5AC is a well-known gastric differentiation marker, which has been frequently used for the classification of stomach cancer. Immunohistochemistry revealed that expression of MUC5AC decreases accompanied with increased malignant property of gastric mucosa, which further suggests the importance of MUC5AC...
Tamás Juhász et al.
Journal of molecular neuroscience : MN, 54(3), 555-573 (2014-08-13)
Presence of the pituitary adenylate cyclase-activating polypeptide (PACAP) signalling has been proved in various peripheral tissues. PACAP can activate protein kinase A (PKA) signalling via binding to pituitary adenylate cyclase-activating polypeptide type I receptor (PAC1), vasoactive intestinal polypeptide receptor (VPAC)...

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。