AV33000

Sigma-Aldrich

Anti-LDB2 (AB1) antibody produced in rabbit

IgG fraction of antiserum

别名:
Anti-LIM domain binding 2
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

43 kDa

species reactivity

guinea pig, mouse, horse, human, dog, rabbit, rat

concentration

0.5 mg - 1 mg/mL

application(s)

western blot: suitable

conjugate

unconjugated

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LDB2(9079)

Immunogen

Synthetic peptide directed towards the N terminal region of human LDB2

Biochem/physiol Actions

LDB2 belongs to the LDB family. LDB2 binds to the LIM domain of a wide variety of LIM domain-containing transcription factors. LIM domains are required for both inhibitory effects on LIM homeodomain transcription factors and synergistic transcriptional activation events. The inhibitory actions of the LIM domain can often be overcome by the LIM co-regulators known as CLIM2, LDB2 and NLI. LIM homeoproteins and CLIMs are involved in a variety of developmental processes.

Sequence

Synthetic peptide located within the following region: DDATLTLSFCLEDGPKRYTIGRTLIPRYFSTVFEGGVTDLYYILKHSKES

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

分析证书
原产地证书 (CofO)

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。