AV34652

Sigma-Aldrich

Anti-KLHL13 antibody produced in rabbit

affinity isolated antibody

别名:
Anti-Kelch-like 13 (Drosophila)
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

74 kDa

species reactivity

human, bovine, horse, mouse, rabbit, guinea pig, rat, dog

concentration

0.5 mg - 1 mg/mL

application(s)

immunohistochemistry: suitable
western blot: suitable

conjugate

unconjugated

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KLHL13(90293)

General description

Kelch-like 13 (KLHL13) belongs to a family of kelch repeat domain containing proteins. It is a component of the BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex. This complex binds to Aurora B and regulates cytokinesis and mitosis. Human KLHL13 is a 73 kDa protein.

Immunogen

Synthetic peptide directed towards the N terminal region of human KLHL13

Biochem/physiol Actions

KLHL13 contains 6 Kelch repeats and 1 BTB (POZ) domain. The function of this protein remains unknown

Sequence

Synthetic peptide located within the following region: KTSSPAIWKFPVPVLKTSRSTPLSPAYISLVEEEDQHMKLSLGGSEMGLS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

分析证书
原产地证书 (CofO)
Izabela Sumara et al.
Developmental cell, 12(6), 887-900 (2007-06-05)
Faithful cell-cycle progression is tightly controlled by the ubiquitin-proteasome system. Here we identify a human Cullin 3-based E3 ligase (Cul3) which is essential for mitotic division. In a complex with the substrate-specific adaptors KLHL9 and KLHL13, Cul3 is required for...
Izabela Sumara et al.
Cell cycle (Georgetown, Tex.), 6(24), 3004-3010 (2007-12-14)
The spindle assembly checkpoint (SAC) is a mechanism that prevents premature chromosome segregation in anaphase before all chromosomes are correctly attached to the mitotic spindle. Errors in chromosome segregation lead to aneuploidy, which may be causally involved in tumorgenesis. Kinetochore...

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。