AV50621

Sigma-Aldrich

Anti-TIPIN (AB1) antibody produced in rabbit

affinity isolated antibody

别名:
Anti-Timeless interacting protein, Anti-FLJ20516
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

34 kDa

species reactivity

bovine, dog, pig, human, mouse, rat

concentration

0.5 mg - 1 mg/mL

application(s)

western blot: suitable

conjugate

unconjugated

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TIPIN(54962)

General description

TIPIN forms a complex with TIMELESS and regulates the stabilization of replication forks. This complex facilitates the cohesion of sister chromatids. It is also involved in Chk1 phosphorylation by ATR during genotoxic stress response.
Rabbit Anti-TIPIN antibody recognizes canine, bovine, human, mouse, and rat TIPIN.

Immunogen

Synthetic peptide directed towards the middle region of human TIPIN

Application

Rabbit Anti-TIPIN antibody is suitable for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

TIPIN belongs to the CSM3 family. TIPIN is required for normal progression of S-phase. It is important for cell survival after DNA damage or replication stress. TIPIN may be specifically required for the ATR-CHK1 pathway in the replication checkpoint induced by hydroxyurea or ultraviolet light.

Sequence

Synthetic peptide located within the following region: SESEMFASELSRSLTEEQQQRIERNKQLALERRQAKLLSNSQTLGNDMLM

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

分析证书
原产地证书 (CofO)
Adam R Leman et al.
Journal of cell science, 123(Pt 5), 660-670 (2010-02-04)
The Timeless-Tipin protein complex has been reported to be important for replication checkpoint and normal DNA replication processes. However, the precise mechanisms by which Timeless-Tipin preserves genomic integrity are largely unclear. Here, we describe the roles of Timeless-Tipin in replication...
Michael G Kemp et al.
The Journal of biological chemistry, 285(22), 16562-16571 (2010-03-18)
Mammalian Timeless is a multifunctional protein that performs essential roles in the circadian clock, chromosome cohesion, DNA replication fork protection, and DNA replication/DNA damage checkpoint pathways. The human Timeless exists in a tight complex with a smaller protein called Tipin...

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。