Merck
BAF134720000
Bel-Art® Write-on Tape Dispenser
Benchtop Model, Multi-Roll
登录查看公司和协议定价
别名:
label tape dispenser, tape cutter
NACRES:
NB.35
制造商/商品名称
Bel-Art F13472-0000
长度 × 宽度 × 高度
30.2 cm × 14.3 cm × 12.1 cm
正在寻找类似产品? Visit 产品对比指南
相关类别
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
BAF134730000BAF134730001BAF134830100
Bel-Art® Write-on™ Label Tape roll size 40 yard, white, width 1 in., pk of 3
BAF134830100
Bel-Art® Write-on Label Tape
L × W × H
30.2 cm × 14.3 cm × 12.1 cm
L × W × H
30.2 cm × 14.3 cm × 12.1 cm
L × W × H
-
L × W × H
-
一般描述
Dispenser holds up to 7 rolls of 1/2" wide labeling tape with a 25mm or 75mm (1" and 3") core diameter. ′Write-On Label Tape is available in 5 colors and 3 widths, 1/2, 3/4 and 1". Differing widths and diameter tapes can be combined on the dispenser. As tape is progressed and severed on the serrated edge, a new section of tape is situated on a platform, ready to make a new label. Includes 3 plastic separating discs and molded-in pen holders 
特点和优势
New piece of label tape is always situated on the flat, smooth writing platform
Load with different color and width label tape to customize to your needs
Can be wall mounted to conserve bench space
其他说明
The Write-On Bench top Tape Dispenser comes with a plastic coated, weighted steel base with rubber feet to avoid slipping on the bench top. The jagged cutting edge is steel for long-term use.
法律信息
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Write-on is a trademark of SP Scienceware, a division of SP
分析证书(COA)
已有该产品?
为方便起见,与您过往购买产品相关的文件已保存在文档库中。
访问文档库
难以找到您所需的产品或批次号码?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA
其他客户在看
Slide 1 of 6
1 of 6
Labeling tape white, W × L 3/4 in. × 500 in.
Z768294
Labeling tape
Bel-Art® Write-on™ Label Tape roll size 40 yard, orange, width 1 in., pk of 3
BAF134880100
Bel-Art® Write-on Label Tape
Bel-Art® Write-on™ Label Tape roll size 40 yard, green, width 1 in., pk of 3
BAF134860100
Bel-Art® Write-on Label Tape
LabJaws™ multi-rod connector nickel plated
Z693812
LabJaws multi-rod connector