Merck
BAF134800050
Bel-Art® Write-on Label Tape
roll size 15 yard, white, width 1/2 in., pk of 6
登录查看公司和协议定价
别名:
labelling tape, moisture resistant label tape, solvent resistant label tape
NACRES:
NB.35
物料
white
包装
pk of 6
manufacturer/tradename
Bel-Art F13480-0050
卷尺寸
15 yard
宽度
1/2 in.
12.7 mm
正在寻找类似产品? Visit 产品对比指南
相关类别
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
BAF134830050BAF134900075BAF134830075
vibrant-m
BAF134800050
Bel-Art® Write-on Label Tape
vibrant-m
BAF134830050
Bel-Art® Write-on Label Tape
vibrant-m
BAF134900075
Bel-Art® Write-on Label Tape
vibrant-m
BAF134830075
Bel-Art® Write-on Label Tape
manufacturer/tradename
Bel-Art F13480-0050
manufacturer/tradename
Bel-Art F13483-0050
manufacturer/tradename
Bel-Art F13490-0075
manufacturer/tradename
Bel-Art F13483-0075
packaging
pk of 6
packaging
pk of 6
packaging
pk of 4
packaging
pk of 4
roll size
15 yard
roll size
40 yard
roll size
60 yard
roll size
40 yard
width
1/2 in.
width
1/2 in. , 12.7 mm
width
19.05 mm , 3/4 in.
width
19.05 mm , 3/4 in.
一般描述
Select from six tape colors that correspond with familiar hazard color codes to highlight safety information written on the labels. Use yellow for chemical reactivity, green for safety labels, blue indicates health hazards, red alerts to flammability; orange indicates a biohazard and white can describe a specific hazard. The tape is custom manufactured from proprietary materials that resist moisture and most solvents. Will adhere to PTFE fluoropolymer resin covered materials and any clean surface. A maximum temperature of 247 degrees C for 30 minutes can be endured.
特点和优势
  • No sticky residue lingers when tape is stripped off
  • Marks made with pencil, ball-point pen or solvent ink marker survive most laboratory conditions
法律信息
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Write-on is a trademark of SP Scienceware, a division of SP
分析证书(COA)
已有该产品?
为方便起见,与您过往购买产品相关的文件已保存在文档库中。
访问文档库
难以找到您所需的产品或批次号码?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA
其他客户在看
Slide 1 of 8
1 of 8
vibrant-m
BAF134620300
Bel-Art® Write-on Label Tape
Sigma label tape white
Sigma-Aldrich
L8394
Sigma label tape
Bel-Art® Write-on™ Label Tape roll size 60 yard, white, width 1/2 in., pk of 6
BAF134900050
Bel-Art® Write-on Label Tape
Bel-Art® Write-on™ Label Tape roll size 40 yard, yellow, width 1 in., pk of 3
BAF134850100
Bel-Art® Write-on Label Tape
Bel-Art® Write-on™ Label Tape roll size 40 yard, red, width 1/2 in., pk of 6
BAF134840050
Bel-Art® Write-on Label Tape
Wash-Off Labels L × W 2.625 in. × 1.00 in., pack of 750 ea (30 labels per sheet)
Z688525
Wash-Off Labels