Merck

HOR human cell line

IHW Number: 9053, 88052053

生物来源

human blood

种族

Japanese

性别

male

生长模式

Suspension

核型

Not specified

形态学

Lymphoblastoid

technique(s)

cell culture | mammalian: suitable

运输

dry ice

正在寻找类似产品? Visit 产品对比指南

比较类似商品

查看完整比较结果

Show Differences

1 of 4

此商品
CB_88052047CB_88052079CB_88052004
HOR human cell line IHW Number: 9053, 88052053

HOR human cell line

PLH human cell line IHW Number: 9047, 88052047

PLH human cell line

EHM human cell line IHW Number: 9080, 88052079

EHM human cell line

JESTHOM human cell line IHW Number: 9004, 88052004

JESTHOM human cell line

ethnicity

Japanese

ethnicity

Scandinavian

ethnicity

German

ethnicity

Scandinavian

gender

male

gender

female

gender

male

gender

male

growth mode

Suspension

growth mode

Suspension

growth mode

Suspension

growth mode

Suspension

karyotype

Not specified

karyotype

Not specified

karyotype

Not specified

karyotype

Not specified

morphology

Lymphoblastoid

morphology

Lymphoblastoid

morphology

Lymphoblastoid

morphology

Lymphoblastoid

一般描述

Consanguineous: No

细胞系描述

This cell line is an Epstein-Barr Virus (EBV) transformed lymphoblastoid line and is part of the Human Leukocyte Antigen (HLA) Typed Collection maintained by the European Collection of Cell Cultures (ECACC). HLA-Typing Data is available for this cell line.

DNA图谱分析

A: 3303
Cw: 1403
B: 44031
HLA DRB1: 1302
HLA DRB3: 301
HLA DQAP: 1.4
HLA DQA1: 102
HLA DQB1: 604
HLA DPB1: 401

血清学分析

HLA A: 33
HLA A ief: 33.2
HLA B: 44
HLA B ief: 44.1
Bw4/bw6: 4
HLA Dw: 19
HLA DR: 13
HLA Dw2: 26
HLA DR52: 52
HLA DQ: 6
HLA DP: 4

培养基

RPMI 1640 (R0883) + 2mM Glutamine (G7513) + 20% Foetal Bovine Serum (FBS) (F2442). Once a growing culture is established the FBS concentration can be reduced to 10%.

传代培养常规

Maintain cultures between 3x105 - 2x106 cells/ml, 5% CO2, 37 °C. Upon resuscitation remove cryoprotectant - centrifuge at 300 x g for 5 minutes and re-suspend the cell pellet in fresh culture medium.

其他说明

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

免责声明

This cell line has restrictions and can only be supplied to academic and not-for-profit organisations. Nucleic acids can be supplied to all researchers including those from commercial organisations.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例 T1503).

示例

T1503
货号
-
25G
包装规格/数量

其它示例:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

请输入1.000309185)

遇到问题?欢迎随时联系我们技术服务 寻求帮助

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich 产品

  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。

  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。

  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。

未找到您寻找的产品?

部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。

索取COA

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门