Merck
C1R human cell line
94050320, IHW Number: 9208
生物来源
human blood
种族
Not specified
性别
female
生长模式
Suspension
核型
Not specified
形态学
Lymphoblastoid
technique(s)
cell culture | mammalian: suitable
运输
dry ice
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
Show Differences
1 of 4
此商品
CB_94050336CB_94050312CB_94050338
C1R human cell line 94050320, IHW Number: 9208
C1R human cell line
CC human cell line 94050336, IHW Number: 9259
CC human cell line
C047 human cell line 94050312, IHW Number: 9265
C047 human cell line
C073 human cell line 94050338, IHW Number: 9261
C073 human cell line
ethnicity
Not specified
ethnicity
African
ethnicity
Spanish Caucasian
ethnicity
Spanish
gender
female
gender
female
gender
not specified
gender
not specified
growth mode
Suspension
growth mode
Suspension
growth mode
Suspension
growth mode
Suspension
karyotype
Not specified
karyotype
Not specified
karyotype
Not specified
karyotype
Not specified
morphology
Lymphoblastoid
morphology
Lymphoblastoid
morphology
Lymphoblastoid
morphology
Lymphoblastoid
一般描述
Consanguineous: No
Homozygous: Yes
细胞系描述
This cell line is an Epstein-Barr Virus (EBV) transformed lymphoblastoid line and is part of the Human Leukocyte Antigen (HLA) Typed Collection maintained by the European Collection of Cell Cultures (ECACC). HLA-Typing Data is available for this cell line.
DNA图谱分析
A: 201
Cw: 401
B: 3503
HLA DRB1: 12
HLA DQB1: 03
血清学分析
HLA C: 4 -
HLA DR: 12
HLA DR52: 52
HLA DQ: 7
培养基
RPMI 1640 (R0883) + 2mM Glutamine (G7513) + 20% Foetal Bovine Serum (FBS) (F2442). Once a growing culture is established the FBS concentration can be reduced to 10%.
传代培养常规
Maintain cultures between 3x105 - 2x106 cells/ml, 5% CO2, 37 °C. Upon resuscitation remove cryoprotectant - centrifuge at 300 x g for 5 minutes and re-suspend the cell pellet in fresh culture medium.
其他说明
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
免责声明
This cell line has restrictions and can only be supplied to academic and not-for-profit organisations. Nucleic acids can be supplied to all researchers including those from commercial organisations.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例 T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
请输入1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA