Merck
All Photos(1)
CLS6795420KIT
Corning® 带有数字显示屏和外部温度探头的加热板搅拌器套件
AC/DC input 120 V AC, 60 Hz, L × W 5 in. × 7 in.
Synonym(s):
magnetic stirrer, magnetic stirrers, stirrers
manufacturer/tradename
Corning
availability
not available in EU
长度 × 宽度
5 in. × 7 in.
Looking for similar products? Visit Product Comparison Guide
一般描述
该Corning套件含有一个带有5" x 7" (12.7 x 17.8 cm)的微晶玻璃顶部的搅拌热板,并具有数字温度和搅拌速度显示。该套件包括一个外部温度控制器(CLS6795PR)、两个9"(22.9 cm)支撑杆(CLS440129)[可连接在一起(18")]、以及搅拌棒取栓器。外部温度控制器和支撑杆可以单独购买,也可以作为附件套件(CLS6795KIT)的一部分购买。它设计为以120V/60Hz的电压运行,并带有标准的美国三爪插头,可用于北美和其他电气要求类似的地区。2年保修期;UL和cUL认证
特点和优势
性能特点:
  • 微处理器可在5°C(如果环境温度为0°C或更低)到550°C的范围内保持一致且可重复的温度设置。
  • 数字LED温度显示可以以5°C为增量进行调节,并闪烁直至达到设定温度。
  • 外部温度控制器真正控制容器内的温度,而无需密切监控液体温度。
  • 独家的闭环搅拌控制可监控和调节60-1150 RPM的搅拌速度,适用于水溶液、粘稠或半固体溶液。
  • 更大、更坚固的磁体改善了搅拌棒的耦合,并结合了自动斜升速度调节功能,从而最大限度地减少了去耦。
安全特性
  • 当顶板温度超过60°C时,即使关闭了加热控制,明亮的LED HOT TOP图标也会亮起。
  • 如果设备过热,独立的温度传感器则会切断电源。
  • 当控制器不在液体中时,外部温度控制器将关闭设备。
  • 两件式铸造设计可将溢出物远离电子设备,并包含一个用于9”至18"(22.9至45.7 cm)支撑杆的内置支架。
法律信息
Corning is a registered trademark of Corning, Inc.
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service