Merck

EHU063341

Sigma-Aldrich

MISSION® esiRNA

targeting human DBNDD1

登录to View Organizational & Contract Pricing

NACRES:
NA.51

描述

Powered by Eupheria Biotech

产品线

MISSION®

形式

lyophilized powder

esiRNA cDNA靶序列

TGCGTGACAGACAAGTCCATTCCCTCCTTAGCTCAGAACACCAAATATCACCAGACTGCCTAAGAGACTTGATGACACCTCCCGGAATGCTCTCGGGGTGGGGTTCACCTCTCCTTGTCCTGCACCCACTGCTAGGCCACATTCTCGTTTCTGCTCACATCCCATTGCCCGGCTACAAGGCCTGCCCACGGCCCTTAAACTTGCTGGGCAGGTTTGGAGCCCATGGGACCCCGTGGGTCTCTGTCCAGGAGCAGCAGAGGAGGCTGACAGGCCCTGCTCCCTCTGCTCTGGGGGTGTCTGGGAGCCCCAGCTCACACCCTCCCAATGCTTATATGCTGAAGCTCACAGAATGGGCTTCTTGCCTGACAGCAAGTCAAAGAATGAGTTTAATATCAAAGTGTAAGCTTACTTTCCATCCCCAAGCCAGCCTGCCCCCTGCCCCATTTCCCATGAGCACACTTCTGGGGAAGGAAA

基因组数据库 |人类登记号

NCBI登记号

运输

ambient

储存温度

−20°C

Gene Information

比较类似商品

查看完整比较结果

显示差异

1 of 4

此商品
EHU042641EHU017351EHU140261
MISSION® esiRNA targeting human DBNDD1

Sigma-Aldrich

EHU063341

MISSION® esiRNA

MISSION® esiRNA targeting human DCTPP1

Sigma-Aldrich

EHU042641

MISSION® esiRNA

MISSION® esiRNA targeting human NOL9

Sigma-Aldrich

EHU017351

MISSION® esiRNA

MISSION® esiRNA targeting human TRIR

Sigma-Aldrich

EHU140261

MISSION® esiRNA

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

esiRNA cDNA target sequence

TGCGTGACAGACAAGTCCATTCCCTCCTTAGCTCAGAACACCAAATATCACCAGACTGCCTAAGAGACTTGATGACACCTCCCGGAATGCTCTCGGGGTGGGGTTCACCTCTCCTTGTCCTGCACCCACTGCTAGGCCACATTCTCGTTTCTGCTCACATCCCATTGCCCGGCTACAAGGCCTGCCCACGGCCCTTAAACTTGCTGGGCAGGTTTGGAGCCCATGGGACCCCGTGGGTCTCTGTCCAGGAGCAGCAGAGGAGGCTGACAGGCCCTGCTCCCTCTGCTCTGGGGGTGTCTGGGAGCCCCAGCTCACACCCTCCCAATGCTTATATGCTGAAGCTCACAGAATGGGCTTCTTGCCTGACAGCAAGTCAAAGAATGAGTTTAATATCAAAGTGTAAGCTTACTTTCCATCCCCAAGCCAGCCTGCCCCCTGCCCCATTTCCCATGAGCACACTTCTGGGGAAGGAAA

esiRNA cDNA target sequence

AGCAGTGCTCTCCAAAATGGACATCAACCGGCGACGCTACCCAGCCCATCTGGCCCGCAGCTCTTCCCGCAAGTATACAGAATTGCCCCATGGGGCCATCTCTGAAGACCAGGCTGTGGGGCCTGCGGACATTCCCTGTGACTCCACAGGCCAGACCTCAACCTAGAAAGATGGCCACAGGACTTGCAACTCAGGGTGGTGTCTGAAGAGCAGAGAGTGGCCTGGCCCTGGAGCCTTTTTCTAGTCTTTTCAGAATAGATCATGGGCCTGAGGCCTCCACTTCTTGAGGTCTGAGGCCCAGCAGCCTCTAGAAGGTAGCCTCCTGGTGTTTGTTCTCCCAGTAAAATGGTTTTGGGCGATAACTTCTAGATTATTCCTGGATGGCCAGGGAGGCTCTCTGTCTCAGCAGGTGATGACGGGGGTACCAGGGGTGCCTCTGAGACCCATTCTCGTGTTTCCCTGT

esiRNA cDNA target sequence

TCCCAGGATGTTGGAAAGTCAACATTTAATAGATACCTGATTAACCATTTGTTAAATAGTCTTCCCTGCGTTGACTATTTGGAATGTGATCTGGGACAGACAGAATTTACCCCTCCTGGTTGCATTTCTTTGCTTAATATTACAGAACCAGTTCTGGGACCACCTTTCACTCACCTGAGGACTCCACAGAAGATGGTATATTATGGGAAACCTTCTTGTAAAAACAACTATGAGAATTATATTGACATAGTGAAATATGTGTTCAGCGCTTACAAGAGAGAGTCCCCTCTCATCGTCAACACTATGGGATGGGTTTCAGACCAGGGGCTCCTGCTTCTCATTGATCTGATCCGATTGCTGTCTCCCAGCCACGTGGTTCAGTTCCGCTCTGACCACAGTAAATATATGCCAGACCTTACCCCGCAGTATGTAGATGACATGGATGGCTTGTACACAAAAAGCAAGACCAAGATGAGAAATCGACGTTTCAGACTCGCAGCATTTG

esiRNA cDNA target sequence

GAAGCAGAAGACGGAGGATGAGGTATTAACAAGTAAAGGTGACGCGTGGGCCAAGTACATGGCAGAAGTGAAAAAGTACAAAGCTCACCAGTGCGGTGACGATGATAAAACTCGGCCCCTGGTGAAATGACGCCCCTCCCCCACCTGCCCATGGCCTGGGACTCTCTGCGATGTACATAACTATTTAATGCAGCGGCAGCGGCGACAGCCTTCCCTGAGAGGACTTAAAAGCAGAAGGAAACCGAGATGCTTCCCGCAGCCGTGGACGATTCTCCAGGACTCTTTTTTTACCTTGAGCACTTGCCTCGTGAGACTTCATAGAACAGTGGTTTACTGTCCCCCCCTTCTCACCTCCTCATTCTCTCTGGCTCTTTCTGTCTTCCTCTTCTCACCCTCCTCCCT

Ensembl | human accession no.

ENSG00000003249

Ensembl | human accession no.

ENSG00000179958

Ensembl | human accession no.

ENSG00000162408

Ensembl | human accession no.

ENSG00000123144

NCBI accession no.

NM_024043

NCBI accession no.

NM_024096

NCBI accession no.

NM_024654

NCBI accession no.

NM_024038

一般描述

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

法律信息

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

储存分类代码

10 - Combustible liquids

闪点(°F)

Not applicable

闪点(°C)

Not applicable


分析证书(COA)

输入产品批号来搜索 分析证书(COA) 。批号可以在产品标签上"批“ (Lot或Batch)字后找到。

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).

示例

T1503
货号
-
25G
包装规格/数量

其它示例:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

输入内容 1.000309185)

遇到问题?欢迎随时联系我们技术服务 寻求帮助

批号可以在产品标签上"批“ (Lot或Batch)字后面找到。

Aldrich 产品

  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。

  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。

  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。

未找到您寻找的产品?

部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。

索取COA

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门