EP1110
Sigma-Aldrich
Sigma-Aldrich® MSMINIDUO buffer saver blocks
2 blocks per package
NACRES:
NC.01
Application
Physically reduce the volume of the gel chamber by up to 100 mL. Package of 2.
Legal Information
Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC