Merck
HS120576
Premium Plus Slide Box
to hold, 100 slide(s), translucent blue acrylonitrile-butadiene-styrene (ABS)
登录查看公司和协议定价
物料
polypropylene
translucent blue acrylonitrile-butadiene-styrene (ABS)
特点
to hold 100 slide(s)
包装
pack of 5 ea
制造商/商品名称
Heathrow Scientific 120576
长度 × 宽度 × 高度
208.3 mm × 195.6 mm × 33 mm
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
HS120593HS120577HS120578
vibrant-m
HS120576
Premium Plus Slide Box
vibrant-m
HS120593
Premium Plus Slide Box
vibrant-m
HS120577
Premium Plus Slide Box
vibrant-m
HS120578
Premium Plus Slide Box
manufacturer/tradename
Heathrow Scientific 120576
manufacturer/tradename
Heathrow Scientific 120593
manufacturer/tradename
Heathrow Scientific 120577
manufacturer/tradename
Heathrow Scientific 120578
material
polypropylene , translucent blue acrylonitrile-butadiene-styrene (ABS)
material
translucent assorted colors acrylonitrile-butadiene-styrene (ABS)
material
polypropylene , translucent orange acrylonitrile-butadiene-styrene (ABS)
material
translucent green acrylonitrile-butadiene-styrene (ABS)
packaging
pack of 5 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of HS120578
L × W × H
208.3 mm × 195.6 mm × 33 mm
L × W × H
-
L × W × H
208.3 mm × 195.6 mm × 33 mm
L × W × H
208.3 mm × 195.6 mm × 33 mm
一般描述
The Premium Plus Slide Box′s modern design for daily use is stylish with a wave surface texture. The slide box is easy to hold,increasing safety of sample transport with an EVA foam lined bottom of the box helping to hold slides securely in placeand protect against damage. Its unique side storage can accommodate a pen, desiccant or other small items needed for working.
特点和优势
  • Made from ABS and Polypropylene makes the boxes robust and eliminates potential rusting of metal parts
  • Inventory card is removable and replaceable for trouble-free writing on flat surfaces.
  • Hinged lid smoothly opens 180° sitting flat on surfaces and prevents misplacement
  • EVA foam lines the bottom of the box helping to hold slides securely in place and protect against damage
  • Multiple labeling areas, a large area on the lid of the box with “project” and “date” molded in and the back of the index card can be used as the lid is transparent
  • Vibrant translucent colors are easy to identify and enable coding options
  • Stylish wave surface texture is easy to hold, increasing safety of sample transport
  • Stackable, molded feet allow secure horizontal stacking of multiple units for safe transport or storing
  • Flexible storage, the units can be stored vertically or horizontally helping to efficiently use space available
  • Unique side storage can accommodate a pen, desiccant or other small items needed for working
  • Holds 100 - 26 x 76 mm slides
  • Patent Pending
分析证书(COA)
已有该产品?
为方便起见,与您过往购买产品相关的文件已保存在文档库中。
访问文档库
难以找到您所需的产品或批次号码?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA