Merck
All Photos(3)

MAK096

Sigma-Aldrich

D-甘露醇检测试剂盒

sufficient for 100 colorimetric tests

NACRES:
NA.84

用途

sufficient for 100 colorimetric tests

detection method

colorimetric

储存温度

2-8°C

Related Categories

一般描述

甘露醇是一种常见的糖醇(多元醇),由许多生物体包括细菌、真菌和植物产生。D-甘露醇作为碳储存化合物、渗透压调节剂和抗氧化剂,通过保护对抗反应性氧化物质 (ROS)。甘露醇是食品生产中常用的低热量甜味剂。糖醇 L-阿拉伯糖醇也作为本试验的底物;因此,本试剂盒可用于测定 L-阿拉伯糖醇。

应用

D -甘露醇比色测定试剂盒已用于评估样品中的细胞内甘露醇含量。

适用性

适用于植物组织、真菌、细菌、食品、果汁、其他饮料等中 D-甘露醇/L-阿拉伯糖醇的检测。

原理

D -甘露醇比色分析试剂盒为测定各种样品(包括水果和果汁)中的 D -甘露醇提供了一种简单直接的程序。通过偶联酶测定法测定 D -甘露醇浓度,其产生与存在的 D-甘露醇成比例的比色 (450 nm) 产物。

储存分类代码

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

(enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service