OGS1892

Sigma-Aldrich

PSF-TEF1-COOH-FXA-MBP - C-TERMINAL MBP TAG YEAST PLASMID

plasmid vector for molecular cloning

别名:
expression vector, molecular cloning vector, vector, snapfast vector, plasmid vector, cloning vector, plasmid
NACRES:
NA.85

form

buffered aqueous solution

mol wt

size 7833 bp

conjugate

maltose-binding protein (MBP) tagged

Origin of replication

2Micron
pUC (500 copies)

Peptide cleavage

Fxa

Peptide tag location

C-terminal

Promoter

Promoter name: TEF1
Promoter activity: constitutive
Promoter type: yeast

bacteria selection

kanamycin

reporter gene

none

shipped in

ambient

storage temp.

−20°C

yeast selection

uracil

General description

This plasmid is designed to express tagged proteins in yeast cells (Saccharomyces cerevisiae). The plasmid contains an auxotrophic Uracil selection expression cassette (URA3) that allows for the positive selection of yeast that are deficient in the URA3 gene (YEL021W). This is typically achieved by growing the yeast in minimal media that is reconstituted with the essential amino acids and nucleotides but excluding Uracil.

About the Peptide Tag:This plasmid contains a c-terminal Maltose Binding Protein (MBP) affinity tag that can be fused to a gene of interest to allow protein detection and/or purification. The sequence of the tag is: EEGKLVIWINGDKGYNGLAEVGKKFEKDTGIKVTVEHPDKLEEKFPQVAATGDGPDIIFWAHDRFGGYAQSGLLAEITPDKAFQDKLYPFTWDAVRYNGKLIAYPIAVEALSLIYNKDLLPNPPKTWEEIPALDKELKAKGKSALMFNLQEPYFTWPLIAADGGYAFKYENGKYDIKDVGVDNAGAKAGLTFLVDLIKNKHMNADTDYSIAEAAFNKGETAMTINGPWAWSNIDTSKVNYGVTVLPTFKGQPSKPFVGVLSAGINAASPNKELAKEFLENYLLTDEGLEAVNKDKPLGAVALKSYEEELAKDPRIAATMENAQKGEIMPNIPQMSAFWYAVRTAVINAASGRQTVDEALKDAQTNSSS.

About the Cleavage Tag:This plasmid also encodes a protease cleavage site that is designed to be positioned between your gene of interest and the tag to allow the removal of the tag following protein purification or isolation. This plasmid contains a FXa cleavage tag. The protein sequence of the cleavage tag is: IEGR. It cleaves after the Arg residue but can also cleave less frequently at secondary basic sites. Its most common secondary cleave site is between the Gly and Arg residues although the frequency of these events is protein specific.

Promoter Expression Level: This plasmid contains the Yeast Elongation Factor Alpha promoter (TEF1). This is the strongest of the yeast promoters that we sell. It is a constitutive promoter and requires no induction. If you are interested in weaker promoters levels than we also stock plasmids that contain the following promoters in order of decreasing strength TPI (strong), ADHI (medium), STE5 (weak). We also stock Galactose inducible promoter plasmids if inducible expression is required. Please contact us for further information.

Sequence

Please select the file type you require. For reference most cloning programs will import a .gb (Genbank) file and will show all of the plasmids features automatically when downloaded and imported.

Genebank Vector Sequence File

FASTA Vector Sequence File

Full Plasmid Map

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxfordgenetics.com.

Other Notes

Looking for more vector options to move your experiments forward faster? Consider a custom cloning vector designed and built by Oxford Genetics. Find out more at Oxford Genetics - Sigma's partner for cloning and expression vectors for molecular biology and synthetic biology applications.

Legal Information

Oxford Genetics is a trademark of Oxford Genetics Ltd

RIDADR

NONH for all modes of transport

分析证书
原产地证书 (CofO)
Alexander C Cerny et al.
PLoS genetics, 11(10), e1005578-e1005578 (2015-10-29)
Recycling of signaling proteins is a common phenomenon in diverse signaling pathways. In photoreceptors of Drosophila, light absorption by rhodopsin triggers a phospholipase Cβ-mediated opening of the ion channels transient receptor potential (TRP) and TRP-like (TRPL) and generates the visual...
Diana Romero et al.
Carcinogenesis, 37(1), 18-29 (2015-10-28)
Dickkopf-3 (Dkk-3) is a secreted protein whose expression is downregulated in many types of cancer. Endogenous Dkk-3 is required for formation of acini in 3D cultures of prostate epithelial cells, where it inhibits transforming growth factor (TGF)-β/Smad signaling. Here, we...
Geoffrey M Lynn et al.
Nature biotechnology, 33(11), 1201-1210 (2015-10-27)
The efficacy of vaccine adjuvants such as Toll-like receptor agonists (TLRa) can be improved through formulation and delivery approaches. Here, we attached small molecule TLR-7/8a to polymer scaffolds (polymer-TLR-7/8a) and evaluated how different physicochemical properties of the TLR-7/8a and polymer...
Jin-Gyoung Jung et al.
PLoS genetics, 10(10), e1004751-e1004751 (2014-10-31)
The Notch3 signaling pathway is thought to play a critical role in cancer development, as evidenced by the Notch3 amplification and rearrangement observed in human cancers. However, the molecular mechanism by which Notch3 signaling contributes to tumorigenesis is largely unknown....

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。