OGS3048

Sigma-Aldrich

PSF-OXB20-NH2-OMPA-V5-TEV - OMPA SECRETION AND V5 TAG PLASMID

plasmid vector for molecular cloning

别名:
expression vector, molecular cloning vector, vector, snapfast vector, plasmid vector, cloning vector, plasmid
NACRES:
NA.85

form

buffered aqueous solution

mol wt

size 3974 bp

conjugate

V5 tagged

Origin of replication

pUC (500 copies)

Peptide cleavage

TEV

Peptide tag location

N-terminal

Promoter

Promoter name: OXB20
Promoter activity: constitutive
Promoter type: bacterial

Secretion signal

OmpA

bacteria selection

kanamycin

reporter gene

none

shipped in

ambient

storage temp.

−20°C

General description

This plasmid is designed to express tagged proteins in E. coli. The plasmid contains a constitutive promoter (OXB20) derived from the region upstream of the E. coli RecA gene. It does not require induction or any additional components for activity. It is the strongest of the bacterial promoters that we provide and this high level of expression can cause expression problems with some proteins with poor solubility. For this reason we sell a range of bacterial promoters with different expression levels (OXB1(low)>OXB20(high)) that can be provided with the peptide tags in this plasmid on request.

About the Peptide Tag:This plasmid contains an Outer Membrane Protein A (OmpA) secretory signal peptide (SP) to allow proteins to be exported from the cytosol. During translocation from the cytosol the signal peptide is removed from the protein by endogenous proteases. This plasmid also contains a secondary V5 protein tag. The sequence of this tag is: GKPIPNPLLGLDSTWe provide a range of dual peptide tag plasmids. This is because some peptide tags provide specific biological properties (e. g., small molecule affinity new epitopes solubility or protein secretion) that are not provided by others.

About the Cleavage Tag: This plasmid also encodes a protease cleavage site that is designed to be positioned between your gene of interest and the tag to allow the removal of the tag following protein purification or isolation. This plasmid contains a TEV cleavage tag. The protein sequence of the cleavage tag is: ENLYFQG. Cleavage occurs between the Glu and Gly residues. TEV is often reported to have better specificity for its recognition site compared to EKT Thrombin or Faxtor Xa.

Promoter Expression Level: This plasmid contains a constitutive bacterial promoter that does not require induction. It is the strongest bacterial promoter we sell and this can cause solubility and expression problems with some proteins. We also offer a range of other bacterial promoters that are compatible with this plasmid and are available on request.

Sequence

Please select the file type you require. For reference most cloning programs will import a .gb (Genbank) file and will show all of the plasmids features automatically when downloaded and imported.

Genebank Vector Sequence File

FASTA Vector Sequence File

Full Plasmid Map

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxfordgenetics.com.

Other Notes

Looking for more vector options to move your experiments forward faster? Consider a custom cloning vector designed and built by Oxford Genetics. Find out more at Oxford Genetics - Sigma's partner for cloning and expression vectors for molecular biology and synthetic biology applications.

Legal Information

Oxford Genetics is a trademark of Oxford Genetics Ltd

RIDADR

NONH for all modes of transport

分析证书
原产地证书 (CofO)
Diana Romero et al.
Carcinogenesis, 37(1), 18-29 (2015-10-28)
Dickkopf-3 (Dkk-3) is a secreted protein whose expression is downregulated in many types of cancer. Endogenous Dkk-3 is required for formation of acini in 3D cultures of prostate epithelial cells, where it inhibits transforming growth factor (TGF)-β/Smad signaling. Here, we...
Geoffrey M Lynn et al.
Nature biotechnology, 33(11), 1201-1210 (2015-10-27)
The efficacy of vaccine adjuvants such as Toll-like receptor agonists (TLRa) can be improved through formulation and delivery approaches. Here, we attached small molecule TLR-7/8a to polymer scaffolds (polymer-TLR-7/8a) and evaluated how different physicochemical properties of the TLR-7/8a and polymer...
Alexander C Cerny et al.
PLoS genetics, 11(10), e1005578-e1005578 (2015-10-29)
Recycling of signaling proteins is a common phenomenon in diverse signaling pathways. In photoreceptors of Drosophila, light absorption by rhodopsin triggers a phospholipase Cβ-mediated opening of the ion channels transient receptor potential (TRP) and TRP-like (TRPL) and generates the visual...
Jin-Gyoung Jung et al.
PLoS genetics, 10(10), e1004751-e1004751 (2014-10-31)
The Notch3 signaling pathway is thought to play a critical role in cancer development, as evidenced by the Notch3 amplification and rearrangement observed in human cancers. However, the molecular mechanism by which Notch3 signaling contributes to tumorigenesis is largely unknown....

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。