PLA0049

Sigma-Aldrich

Goat anti-RFC4 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

别名:
replication factor C (activator 1) 4, RFC 37 kDa subunit, activator 1 37 kDa subunit, replication factor C 37 kDa subunit, activator 1 subunit 4, A1, A1 37 kDa subunit, RF-C 37 kDa subunit, replication factor C (activator 1) 4 (37kD), 37kDa, RFC37
NACRES:
NA.41

Quality Level

biological source

goat

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

application(s)

western blot: 1:500-1:2,500

accession no.

NP_002907.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

goat ... RFC4(5984)

Immunogen

The epitope recognized by PLA0049 maps to a region between residues 300 and 350 of human Replication Factor C subunit 4 using the numbering given in entry NP_002907.1 (GeneID 5984).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

The Replication factor C (RFC) heteropentamer includes RFC1, RFC2, RFC3, RFC4, and RFC5. The RFC heteropentamer is part of the multiprotein DNA replicase that functions to replicate DNA prior to cell division and repair DNA damage. RFC is the clamp loader ATPase associated with the DNA replicase. As the clamp loader, RFC uses ATP to recruit, open, and close the PCNA ring to link it to DNA for replication and repair. RFC4 is the 37 kDa subunit of RFC that is part of the core complex formed with RFC2 and RFC5.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

WGK Germany

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

分析证书
原产地证书 (CofO)

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。