Merck
  • SAB1412527
All Photos(3)

SAB1412527

Sigma-Aldrich

ANTI-SNAI2 antibody produced in mouse

clone 6A7, purified immunoglobulin, buffered aqueous solution

Synonym(s):
MGC10182, SLUG, SLUGH1, SNAI2, WS2D
NACRES:
NA.41

生物来源

mouse

质量水平

偶联物

unconjugated

抗体形式

purified immunoglobulin

antibody product type

primary antibodies

克隆

6A7, monoclonal

形式

buffered aqueous solution

分子量

antigen 33.77 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

同位素/亚型

IgG1κ

NCBI登记号

UniProt登记号

运输

dry ice

储存温度

−20°C

Gene Information

human ... SNAI2(6591)

一般描述

This gene encodes a member of the Snail family of C2H2-type zinc finger transcription factors. The encoded protein acts as a transcriptional repressor that binds to E-box motifs and is also likely to repress E-cadherin transcription in breast carcinoma. This protein is involved in epithelial-mesenchymal transitions and has antiapoptotic activity. Mutations in this gene may be associated with sporatic cases of neural tube defects. (provided by RefSeq)

免疫原

SNAI2 (NP_003059, 97 a.a. ~ 169 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KDHSGSESPISDEEERLQSKLSDPHAIEAEKFQCNLCNKTYSTFSGLAKHKQLHCDAQSRKSFSCKYCDKEYV

外形

Solution in phosphate buffered saline, pH 7.4

免责声明

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

储存分类代码

12 - Non Combustible Liquids

闪点(F)

Not applicable

闪点(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service