SAB2102371

Sigma-Aldrich

Anti-TARDBP (ab1) antibody produced in rabbit

affinity isolated antibody

别名:
Anti-TAR DNA binding protein, Anti-TDP-43
NACRES:
NA.41

生物来源

rabbit

质量水平

100

抗体形式

affinity isolated antibody

antibody product type

primary antibodies

克隆

polyclonal

形式

buffered aqueous solution

分子量

45 kDa

species reactivity

dog, human, bovine, rat, mouse, horse, guinea pig, rabbit

浓度

0.5 mg - 1 mg/mL

应用

immunohistochemistry: suitable
western blot: suitable

偶联物

unconjugated

UniProt登记号

运输

wet ice

储存温度

−20°C

Gene Information

human ... TARDBP(23435)

免疫原

Synthetic peptide directed towards the N terminal region of human TARDBP

生化/生理作用

HIV-1, the causative agent of acquired immunodeficiency syndrome (AIDS), contains an RNA genome that produces a chromosomally integrated DNA during the replicative cycle. Activation of HIV-1 gene expression by the transactivator Tat is dependent on an RNA regulatory element (TAR) located downstream of the transcription initiation site. TARDBP is a transcriptional repressor that binds to chromosomally integrated TAR DNA and represses HIV-1 transcription.HIV-1, the causative agent of acquired immunodeficiency syndrome (AIDS), contains an RNA genome that produces a chromosomally integrated DNA during the replicative cycle. Activation of HIV-1 gene expression by the transactivator Tat is dependent on an RNA regulatory element (TAR) located downstream of the transcription initiation site. The protein encoded by this gene is a transcriptional repressor that binds to chromosomally integrated TAR DNA and represses HIV-1 transcription. In addition, this protein regulates alternate splicing of the CFTR gene. A similar pseudogene is present on chromosome 20. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

序列

Synthetic peptide located within the following region: VYVVNYPKDNKRKMDETDASSAVKVKRAVQKTSDLIVLGLPWKTTEQDLK

外形

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

免责声明

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

storage_class_code

12 - Non Combustible Liquids

WGK Germany

WGK 3

闪点(F)

Not applicable

闪点(C)

Not applicable

分析证书

原产地证书 (CofO)

Jack L Pinkus et al.
Neuromuscular disorders : NMD, 24(7), 611-616 (2014-05-27)
Previous histopathologic studies of sporadic inclusion body myositis (sIBM) identified sarcoplasmic aggregation and myonuclear depletion of the predominantly nuclear heterogeneous nuclear ribonucleoprotein (hnRNP) TDP-43 in sIBM myofibers. Here, we examined sIBM muscle for abnormalities in two other hnRNPs hnRNPA1 and...
Hideaki Tagashira et al.
Biochimica et biophysica acta, 1840(12), 3320-3334 (2014-09-02)
Amyotrophic lateral sclerosis (ALS) is a disease caused by motor neuron degeneration. Recently, a novel SIGMAR1 gene variant (p.E102Q) was discovered in some familial ALS patients. We address mechanisms underlying neurodegeneration caused by the mutation using Neuro2A cells overexpressing σ1R(E102Q)...
Chi-Chen Huang et al.
Journal of cell science, 127(Pt 14), 3024-3038 (2014-05-27)
TDP-43 (also known as TARDBP) is a pathological signature protein of neurodegenerative diseases, with TDP-43 proteinopathies including frontotemporal lobar degeneration (FTLD)-TDP and amyotrophic lateral sclerosis (ALS)-TDP. These TDP-43 proteinopathies are characterized by cytoplasmic insoluble TDP-43-positive aggregates in the diseased cells...

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门