Merck

SAB5701830

Sigma-Aldrich

Anti-Phospho-p53-S376 antibody produced in rabbit

登录查看公司和协议定价

生物来源

rabbit

质量水平

抗体产品类型

primary antibodies

克隆

monoclonal

形式

liquid

种属反应性

human, mouse

浓度

1.4 mg/mL

技术

western blot: 1:500-1:2000

UniProt登记号

运输

wet ice

储存温度

−20°C

靶向翻译后修饰

phosphorylation (pSer376)

基因信息

human ... TP53(7157)

比较类似商品

查看完整比较结果

显示差异

1 of 4

此商品
SAB5701828SAB5701827SAB5701804
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Gene Information

human ... TP53(7157)

Gene Information

human ... TP53(7157)

Gene Information

human ... TP53(7157)

Gene Information

human ... TP53(7157)

clone

monoclonal

clone

monoclonal

clone

monoclonal

clone

polyclonal

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

一般描述

This gene encodes a tumor suppressor protein containing transcriptional activation, DNA binding, and oligomerization domains. The encoded protein responds to diverse cellular stresses to regulate expression of target genes, thereby inducing cell cycle arrest, apoptosis, senescence, DNA repair, or changes in metabolism. Mutations in this gene are associated with a variety of human cancers, including hereditary cancers such as Li-Fraumeni syndrome. Alternative splicing of this gene and the use of alternate promoters result in multiple transcript variants and isoforms. Additional isoforms have also been shown to result from the use of alternate translation initiation codons from identical transcript variants (PMIDs: 12032546, 20937277). [provided by RefSeq, Dec 2016]

免疫原

A phospho specific peptide corresponding to residues surrounding S376 of human p53

外形

Buffer: PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

储存分类代码

12 - Non Combustible Liquids

WGK

WGK 1

闪点(°F)

Not applicable

闪点(°C)

Not applicable


分析证书(COA)

输入产品批号来搜索 分析证书(COA) 。批号可以在产品标签上"批“ (Lot或Batch)字后找到。

已有该产品?

为方便起见,与您过往购买产品相关的文件已保存在文档库中。

访问文档库

难以找到您所需的产品或批次号码?

在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).

示例

T1503
货号
-
25G
包装规格/数量

其它示例:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

输入内容 1.000309185)

遇到问题?欢迎随时联系我们技术服务 寻求帮助

批号可以在产品标签上"批“ (Lot或Batch)字后面找到。

Aldrich 产品

  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。

  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。

  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。

未找到您寻找的产品?

部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。

索取COA

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门