Merck
SRP5271
Sigma-Aldrich
SIRT2, active, His tagged human
recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
别名:
SIR2L, SIR2L2
NACRES:
NA.32
重组
expressed in baculovirus infected Sf9 cells
检测方案
≥70% (SDS-PAGE)
形式
buffered aqueous glycerol solution
specific activity
6120-10580 RLU/min·ng
分子量
~42 kDa
NCBI登记号
运输
dry ice
储存温度
−70°C
Gene Information
human ... SIRT2(22933)
一般描述
SIRT2 is a member of the sirtuin family of proteins which are homologs to the yeast Sir2 protein. Sirtuin family contain a sirtuin core domain and are grouped into four classes with SIRT2 being a member of class I. Inhibition of SIRT2 can lead to neuroprotection in cellular and invertebrate models of Huntington′s disease. Huntington′s disease is characterized by increased sterol synthesis in neuronal cells and this process is reversed by SIRT2 inhibition. SIRT2 can deacetylate lys40 of alpha-tubulin both in vitro and in vivo. Knockdown of SIRT2 via small interfering RNA results in tubulin hyperacetylation.
外形
Supplied in 50 mM sodium phosphate, pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, 25% glycerol.
制备说明
After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.
象形图
Exclamation markHealth hazard
警示用语:
Danger
危险分类
Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2
储存分类代码
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
闪点(F)
Not applicable
闪点(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例 T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
请输入1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA
Sirtuin 2 mutations in human cancers impair its function in genome maintenance.
Head PE
The Journal of Biological Chemistry (2017)