Merck
Z740520
Brady® BMP51 printer kit
AC/DC input 100 - 240 V AC (GB plug)
登录to View Organizational & Contract Pricing
Select a Size
NACRES:
NB.35
特点
Kit includes: BMP51 Printer with battery pack and AC adaptor/battery charger
包装
pkg of 1 kit
manufacturer/tradename
Brady 710898GB
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
Z740522Z664529Z664723
Brady® BMP51 printer kit AC/DC input 100 - 240 V AC (GB plug)
Z740520
Brady® BMP51 printer kit
Brady® BMP51 printer kit AC/DC input 100 - 240 V AC (US plug)
Z740522
Brady® BMP51 printer kit
Brady® Freezerbondz™ labels L × W 31.75 mm × 9.53 mm
Z664529
Brady® Freezerbondz labels
Brady® Freezerbondz™ labels L × W 55.88 mm × 38.10 mm
Z664723
Brady® Freezerbondz labels
feature
Kit includes: BMP51 Printer with battery pack and AC adaptor/battery charger
feature
Kit includes: BMP51 Printer with battery pack and AC adaptor/battery charger
feature
-
feature
-
manufacturer/tradename
Brady 710898GB
manufacturer/tradename
Brady 75447314703
manufacturer/tradename
Brady PTL-112-490
manufacturer/tradename
Brady X-113-490
一般描述
The BMP51 Label Maker is a compact, versatile portable printer with built-in Bluetooth® technology for wireless printing. When used in conjunction with the free Brady Mobile App, you can design and print labels with this printer using your Android phone!
This label maker has a 300 dpi print head and can print professional, durable labels up to 38.10 mm wide. It prints on continuous and die-cut materials at a speed of 25.40 mm per second. With a built-in cutter, easy top-loading cartridges, and 3 different options for battery power, the BMP51 Label Maker is a powerful printer for on-the-job label making.
The BMP51 has a full QWERTY keyboard, for stand alone operation. The BMP51 Label Maker is ideal for electrical, datacom, facility/safety, product and panel manufacturing, and laboratory labelling applications.
Get the job done faster with more confidence.
  • Change supplies in seconds with hassle-free drop-in cartridges
  • Automatic label formatting – no printer calibration!
  • QWERTY style keypad for faster, more accurate typing
  • Battery options for portability (AA batteries, rechargeable Li-Ion battery or AC power)
  • Labels up to 1.5 in. wide
  • Continuous & die-cut label materials
  • Import, store, and print label files from Excel®
  • Brady labels will not fade, smear or fall off – so you only need to label something once
  • Rugged printer design can survive a 4-foot drop onto concrete, harsh environments and extreme temperatures
法律信息
Bluetooth is a registered trademark of Bluetooth SIG, Inc.
Brady is a registered trademark of Brady Worldwide, Inc.
Excel is a registered trademark of Microsoft Corporation
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA