19138-U

Hamilton® needles for RN syringes, blunt tip

needle size 26s ga, needle L 51 mm (2 in.), for use with 2.5μL-100μL syringe, blunt tip (point style 3)

NACRES:
NB.03

物料

blunt tip stainless steel needle

针尺寸

26s ga

描述

blunt tip (stainless steel)

特点

blunt tip (point style 3)

包装

pkg of 6 ea

manufacturer/tradename

Hamilton 7768-01

针长度

51 mm (2 in.)

体积

2.5-100 μL

相容性

for use with 2.5μL-100μL syringe
for use with Rheodyne Injection Valve

正在寻找类似产品? Visit 产品对比指南

一般描述

Hamilton® needles for RN syringes are equipped with 90° blunt tips which have chamfered and polished edges that eliminate damage to the valve′s rotor seal and stator face. This style also can be used for pipetting of liquids.

法律信息

Hamilton is a registered trademark of Hamilton Co.

storage_class_code

13 - Non Combustible Solids

WGK Germany

WGK 3

闪点(F)

Not applicable

闪点(C)

Not applicable

分析证书

原产地证书 (CofO)

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门