67088-U

Supelco

BIOshell A160 Peptide C18, 2.7 μm HPLC Column

2.7 μm particle size, L × I.D. 15 cm × 100 μm

NACRES:
SB.52

Quality Level

material

stainless steel hardware

Agency/Method

suitable for USP L1

description

Shell thickness (0.5 μm)
Solid Core (1.7 μm)

feature

endcapped: no

analyte chemical class(es)

peptides

manufacturer/tradename

BIOshell

extent of labeling

4.6% carbon loading

parameter

600 bar max. pressure
90 °C max. temp.

application(s)

HPLC: suitable
UHPLC: suitable

L × I.D.

15 cm × 100 μm

surface coverage

2.2 μmol/m2

matrix

spherical silica particle platform

matrix active group

C18 (octadecyl) bonding phase, diisobutyloctadecyl

particle size

2.7 μm

pore size

160 Å

surface area

90 m2/g

operating pH range

1 - 8

separation technique

reversed phase

正在寻找类似产品? Visit 产品对比指南

相关类别

General description

BIOshell 160 Å Peptide C18 columns are specifically engineered to provide efficient separation of peptides as well as small proteins. These columns contain advanced Fused-Core particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping. Additionally, the sterically-protected C18 ligands provide extra stability allowing the columns to be used at an extended pH range (2-9) and high temperatures (up to 90 °C). This greatly expands the application range for the separation of biomolecules.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

BIOshell is a trademark of Sigma-Aldrich Co. LLC
技术文章
Development of Simple and Rapid Workflows for Quantitation of Infliximab and Adalimumab in Human Serum by LC-MS/MS
了解更多

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。