技术
gas chromatography (GC): suitable
试验参数
column: γ-DEX 120, 30 m × 0.25 mm I.D., 0.25 μm (24307)
oven: 90 °C, 1 °C/min to 135 °C, 5 °C/min to 240 °C
inj. temp.: 300 °C
detector: FID, 300 °C
carrier gas: helium, 20 cm/sec
injection: 1 μL 100:1 split
sample: 500 μg/mL in methylene chloride
应用
agriculture
chemicals and industrial polymers
forensics and toxicology
法律信息
DEX is a trademark of Sigma-Aldrich Co. LLC
分析物
说明
2-Ethylhexanoic acid
α-BHC
分析色谱柱