Merck
All Photos(1)

ES5220

GreenSep Ethyl Pyridine SFC 5 Micron HPLC Column

5 μm particle size, L × I.D. 15 cm × 3.0 mm

NACRES:
SB.52

物料

stainless steel column

质量水平

100

产品线

GreenSep

manufacturer/tradename

ES Industries 133291-GS-PYE

包装

1 ea of

technique(s)

HPLC: suitable

长度 × 内径

15 cm × 3.0 mm

基质

fully porous particle

基质活性基团

2-ethyl pyridine phase

粒径

5 μm

孔径

120 Å pore size

operating pH

2-8

分离技术

supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

Related Categories

一般描述

The type of chemicals separated on a GreenSep Ethyl Pyridine column would normally require the addition of an amine to the mobile phase as a peak shape modifier; however GreenSep Ethyl Pyridine does not require this addition. Ideal for chemicals that are functionalized with strong amine groups, GreenSep Ethyl Pyridine provides flexibility for the SFC chromatographer with mobile phase composition and fraction collection greatly simplified without the use of amino additives.

推荐产品

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

法律信息

GreenSep is a trademark of ES Industries

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service