ES5345

GreenSep Pyridyl Amide SFC 3 Micron HPLC Column

3 μm particle size, L × I.D. 10 cm × 2.1 mm

NACRES:
SB.52

Quality Level

manufacturer/tradename

ES Industries 122191-GS-PYA

application(s)

HPLC: suitable

L × I.D.

10 cm × 2.1 mm

matrix active group

pyridyl amide phase

particle size

3 μm

pore size

120 Å pore size

separation technique

supercritical fluid (SFC)

正在寻找类似产品? Visit 产品对比指南

General description

The type of chemicals separated on a GreenSep Pyridyl Amide column would normally require the addition of TFA or an amine to the mobile phase as a peak shape modifier; however GreenSep Pyridyl Amide does not require this addition. Ideal for chemicals that contain both basic amine and acidic groups, as shown in the chromatogram below, GreenSep Pyridyl Amide provides flexibility for the SFC chromatographer with mobile phase composition and fraction collection greatly simplified without the use of amino additives.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries
分析证书
原产地证书 (CofO)

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。