ES5577
ChromegaChiral Chiral CCO F4, 3 Micron HPLC Column
3 μm particle size, L × I.D. 10 cm × 3.0 mm
NACRES:
SB.52
manufacturer/tradename
ES Industries 123151-CCO-F4
Quality Level
application(s)
HPLC: suitable
L × I.D.
10 cm × 3.0 mm
particle size
3 μm
pore size
1000 Å pore size
separation technique
chiral
正在寻找类似产品? Visit 产品对比指南
General description
ChromegaChiral CCO F4 is a 4-Fluoro 3-methylphenyl cellulose phase which can be used in SFC or HPLC. The addition of a fluorine atom into a phenyl carbamate cellulose structure is useful in promoting a fluorophilic retention mechanism which provides improved retention for fluorinated compounds. A fluorophilic retention mechanism is particularly useful in medicinal chemistry and drug discovery, where more than a third of newly approved small molecule drugs contain fluorine.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
ChromegaChiral is a trademark of ES Industries