技术规格说明书
产品名称
2,3,4,5,6-五氟溴化苄 99%
货号
101052
产品品牌
ALDRICH
CAS号
1765-40-8
分子量
260.99
测试
规格
APPEARANCE (COLOR)Colorless to Yellow to Brown
APPEARANCE (FORM)Liquid or Solid
REM. ON APPEARANCEMaterial may darken on storage with no loss in purity
PURITY (GC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE