技术规格说明书
产品名称
氧化锆 (IV) 99.99% trace metals basis (purity excludes ~2% HfO2)
货号
204994
产品品牌
ALDRICH
CAS号
1314-23-4
分子量
123.22
测试
规格
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder
PURITY99.99% PURITY BASED ON TRACE METALS ANALYSIS (EXCLUDES ~2% HfO2)
ICP ANALYSISCONFIRMS ZIRCONIUM COMPONENT
METAL TRACE ANALYSIS (ICP)≤ 200 PPM TRACE METALLIC IMPURITIES (excludes Hf content).