技术规格说明书
产品名称
氧化锆 (IV) powder, 5 μm, 99% trace metals basis
货号
230693
产品品牌
ALDRICH
CAS号
1314-23-4
分子量
123.22
测试
规格
APPEARANCEWHITE POWDER
X-RAY DIFFRACTIONCONFORMS TO STANDARD PATTERN.
 AND/OR
ICP ASSAYCONFIRMS ZIRCONIUM COMPONENT.
LOSS ON IGNITION1.5% (MAXIMUM) (1000 DEGREES C, 1 HOUR)
 TYPICALLY CA. 0.5% OR LESS
TRACE ANALYSIS, ICP99% (MINIMUM) AS DEFINED BY DETERMINATION OF
 TRACE METALLIC IMPURITIES. HAFNIUM CONTENT
 2.0% (MAXIMUM)
PARTICLE SIZE<5 MICRON AVERAGE PARTICLE SIZE
 REVISED MAY 16, 1996 JSB