技术规格说明书
产品名称
钛 nanoparticles, dispersion, <100 nm particle size, in mineral oil, 98.5% trace metals basis
货号
513415
产品品牌
ALDRICH
CAS号
7440-32-6
分子量
47.87
测试
规格
APPEARANCEGREY-BLACK TO BLACK DISPERSION
TRACE ANALYSIS, ICP15,000 PPM (MAXIMUM) TRACE METALLIC
 IMPURITIES
ICP ASSAYCONFIRMS TITANIUM COMPONENT.
PURITYPURITY BASED ON TRACE METAL CONTENT.
PARTICLE SIZE100 NM (MAXIMUM) AVERAGE PARTICLE SIZE
 REVISED MAY 15, 2007 RJM