技术规格说明书
产品名称
过氧化氢 溶液 contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
货号
323381
产品品牌
SIGALD
CAS号
7722-84-1
分子量
34.01
测试
规格
STABILIZER150 - 250 PPM ACETANILIDE
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
REDOX TITRATION3.0 WT%-4.0 WT% H2O2 (WITH KMNO4)