技术规格说明书
产品名称
D -(+)-葡萄糖 溶液 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
货号
G8769
产品品牌
SIGMA
CAS号
50-99-7
分子量
180.16
测试
规格
Appearance (Turbidity)Clear
Appearance (Form)Solution
pH3.0 - 4.0
Cell Culture Testing - MTTPass
Cell LineCell Line - Cell Types
SterilityPass
Endotoxin Level<= 0.5 EU/ml
Glucose Concentration405 - 495 g/l