技术规格说明书
产品名称
氢氧化钠 溶液 1.0 N, BioReagent, suitable for cell culture
货号
S2770
产品品牌
SIGMA
CAS号
1310-73-2
分子量
40.00
测试
规格
Appearance (Turbidity)Clear
Appearance (Form)Solution
pH-
Osmolality- mOs/kg
Cell Culture Testing - MTTPass
Cell LineCell Line - Cell Types
Key Element Conc - ICPPass
SterilityPass
Normality by Titration0.9 - 1.1
Not Derived From Animal OriginConforms