技术规格说明书
产品名称
Anti-ZNF205 antibody produced in rabbit IgG fraction of antiserum
货号
av38353
产品品牌
sigma
CAS号
分子量
测试
规格
Concentration0.5-1 mg/ml
ELISANot Verified
Western BlottingAntibody dilution: 1 µg/ml
ImmunohistochemistryNot Verified
ImmunofluorescenceNot Verified
ImmunoprecipitationNot Verified
Flow CytometryNot Verified
Chromatin ImmunoprecipitationNot Verified