技术规格说明书
产品名称
Dipeptidyl Peptidase VII human recombinant, expressed in Sf9 cells
货号
d3321
产品品牌
sigma
CAS号
分子量
测试
规格
units/µg protein≥1500
 Enzyme Activity: One unit will hydrolyze 1
 picomole of Ala-Pro-AMC per minute at pH 7.4 at 25
 deg C
FormulationConforms
 Solution in 10% glycerol containing 25 mM Tris-HCl
 , pH 8.0, 130 mM NaCl, and 0.05% Tween-20.