HK
ZH
应用
产品
服务
支持
Home
Search Results
乙腈EMPROVE
®
ESSENTIAL
(1)
乙腈EMPROVE
®
ESSENTIAL
Synonyms:
乙腈, 甲基氰, 氰甲烷, ACN
Linear Formula:
CH
3
CN
CAS Number:
75-05-8
Molecular Weight:
41.05
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
1.37134
乙腈EMPROVE
®
ESSENTIAL, acetonitrile processed under GMP conditions for use in pharmaceutical production, EMPROVE
®
ESSENTIAL
Expand
View Pricing