返回
MAS 100 NT的带孔盖(300 x 0.6 mm)
别名:
穿孔盖
Products
产品分类
accessories (7)
品牌
Millipore (7)
机构
直径(mm)
相容性
应用
筛选产品
Showing 1-7 of 7
比较
货号
说明
价格
1.09195
MAS 100 NT的带孔盖(300 x 0.6 mm),
300 hole x 0.6mm, Aluminium, Suitable for MAS-100 NT
®
/ MAS-100 VF
®
展开
查看定价
1.09088
MAS 100 NT的带孔盖(300 x 0.6 mm),
400 hole x 0.7mm, Aluminium, for MAS-100 Eco
®
展开
查看定价
1.09189
MAS 100 NT的带孔盖(300 x 0.6 mm),
300 x 0.6 mm, Stainless steel, edge protected for MAS-100 Iso
®
line air sampler
展开
查看定价
1.09213
MAS 100 NT的带孔盖(300 x 0.6 mm),
400 hole x 0.7mm, Aluminium, suitable for portable MAS-100
®
family and non-lockable ICR contact plates
展开
查看定价
1.19363
MAS 100 NT的带孔盖(300 x 0.6 mm),
300 x 0.6 mm, aluminum, edge-protected for MAS-100 NT
®
and MAS-100 VF
®
展开
查看定价
1.19364
MAS 100 NT的带孔盖(300 x 0.6 mm),
400 x 0.7 mm, aluminum, edge-protected MAS-100 Eco
®
展开
查看定价
1191490001
MAS 100 NT的带孔盖(300 x 0.6 mm),
300 hole x 0.6mm, Aluminium, suitable for portable MAS-100 NT
®
, MAS-100 VF
®
and ICRplus contact plates